ENA413170202 Enamine

1-[3-(3-fluorophenyl)-1,2,4-oxadiazol-5-yl]ethyl acetate

  • MDL number MFCD27500809
  • Cas Number : 1803590-04-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153063

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153063
Availability details for
Quantity
  • Availability
  • Add to Cart