ENA413170206 Enamine

2-amino-3-(trimethyl-1H-pyrazol-1-yl)propanoic acid dihydrochloride

  • MDL number MFCD27500812
  • Cas Number : 1803565-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153100

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153100
Availability details for
Quantity
  • Availability
  • Add to Cart