ENA413170207 Enamine

1-(2-methoxyethyl)cyclohexane-1-carbaldehyde

  • MDL number MFCD27920202
  • Cas Number : 1803590-11-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153130

Safety Information

msds https://www.enaminestore.com/catalog/EN300-153130
Availability details for
Quantity
  • Availability
  • Add to Cart