ENA413170210 Enamine

methyl 4-(5-methylthiophen-2-yl)-1H-pyrrole-3-carboxylate

  • MDL number MFCD27500817
  • Cas Number : 1803611-72-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-153198

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153198
Availability details for
Quantity
  • Availability
  • Add to Cart