ENA413170222 Enamine

2-[(5-oxo-4,5-dihydro-1H-1,2,4-triazol-3-yl)sulfanyl]acetic acid

  • MDL number MFCD27500826
  • Cas Number : 141956-66-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153311

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153311
Availability details for
Quantity
  • Availability
  • Add to Cart