ENA413170224 Enamine

N-methyl-3,4,5,6-tetrahydropyridin-2-amine hydrochloride

  • MDL number MFCD27500827
  • Cas Number : 3256-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153326

Safety Information

msds https://www.enaminestore.com/catalog/EN300-153326
Availability details for
Quantity
  • Availability
  • Add to Cart