ENA413170225 Enamine

propan-2-yl N-(piperazine-1-sulfonyl)carbamate hydrochloride

  • MDL number MFCD27500828
  • Cas Number : 1803606-68-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153328

Safety Information

msds https://www.enaminestore.com/catalog/EN300-153328
Availability details for
Quantity
  • Availability
  • Add to Cart