ENA413170226 Enamine

4-bromooxane-4-carbaldehyde

  • MDL number MFCD26936304
  • Cas Number : 1602211-57-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153338

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153338
Availability details for
Quantity
  • Availability
  • Add to Cart