ENA413170229 Enamine

1-(4-methylphenyl)-1H-imidazo[4,5-c]pyridine

  • MDL number MFCD27500832
  • Cas Number : 1803599-43-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153437

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153437
Availability details for
Quantity
  • Availability
  • Add to Cart