ENA413170231 Enamine

2-[1-(4-methylpiperazin-1-yl)cyclohexyl]acetic acid dihydrochloride

  • MDL number MFCD27500835
  • Cas Number : 1797943-35-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153450

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153450
Availability details for
Quantity
  • Availability
  • Add to Cart