ENA413170241 Enamine

[3-(dimethylamino)-1,2,4-oxadiazol-5-yl]methanol

  • MDL number MFCD27500853
  • Cas Number : 1602419-10-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153620

Safety Information

msds https://www.enaminestore.com/catalog/EN300-153620
Availability details for
Quantity
  • Availability
  • Add to Cart