ENA413170242 Enamine

potassium 5-oxo-2,5-dihydro-1,2,4-oxadiazole-3-carboxylate

  • MDL number MFCD27500854
  • Cas Number : 1803582-45-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153621

Safety Information

msds https://www.enaminestore.com/catalog/EN300-153621
Availability details for
Quantity
  • Availability
  • Add to Cart