ENA413170257 Enamine

8-fluoro-6-methylquinoline-3-carbonitrile

  • MDL number MFCD27500874
  • Cas Number : 1597983-90-8
  • Molecular Weight : 186.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 8-fluoro-6-methylquinoline-3-carbonitrile
Inchi Code 1S/C11H7FN2/c1-7-2-9-4-8(5-13)6-14-11(9)10(12)3-7/h2-4,6H,1H3
Inchi Key XPOJHFTTYZWYRA-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-153772

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153772
Availability details for
Quantity
  • Availability
  • Add to Cart