ENA413170260 Enamine

(5-bromo-7-methyl-1-benzofuran-2-yl)methanamine hydrochloride

  • MDL number MFCD27500878
  • Cas Number : 1803592-29-1
  • Molecular Weight : 276.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (5-bromo-7-methylbenzofuran-2-yl)methanamine hydrochloride
Inchi Code 1S/C10H10BrNO.ClH/c1-6-2-8(11)3-7-4-9(5-12)13-10(6)7;/h2-4H,5,12H2,1H3;1H
Inchi Key HWZMUEVWSGJURK-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-153792

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153792
Availability details for
Quantity
  • Availability
  • Add to Cart