ENA413170264 Enamine

benzyl 4-[(1H-1,2,4-triazol-1-yl)methyl]piperidine-1-carboxylate

  • MDL number MFCD27500882
  • Cas Number : 1803580-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153846

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153846
Availability details for
Quantity
  • Availability
  • Add to Cart