ENA413170268 Enamine

2-(butyldimethylazaniumyl)acetate

  • MDL number MFCD27500888
  • Cas Number : 228711-03-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153907

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153907
Availability details for
Quantity
  • Availability
  • Add to Cart