ENA413170274 Enamine

2-chloro-6-fluoro-8-nitroquinoline

  • MDL number MFCD27920206
  • Cas Number : 1803580-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153967

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153967
Availability details for
Quantity
  • Availability
  • Add to Cart