ENA413170282 Enamine

4-(methylamino)-1,2-dihydropyrimidin-2-one hydrochloride

  • MDL number MFCD27500908
  • Cas Number : 1195687-26-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154133

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154133
Availability details for
Quantity
  • Availability
  • Add to Cart