ENA413170287 Enamine

1-[6-(2-chloroacetyl)-1,2,3,4-tetrahydroisoquinolin-2-yl]-2,2,2-trifluoroethan-1-one

  • MDL number MFCD27920208
  • Cas Number : 1803611-75-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-154350

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154350
Availability details for
Quantity
  • Availability
  • Add to Cart