ENA413170295 Enamine

methyl 6-(4-fluorophenyl)-6-oxohexanoate

  • MDL number MFCD27500923
  • Cas Number : 342636-35-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154614

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154614
Availability details for
Quantity
  • Availability
  • Add to Cart