ENA413170299 Enamine

3-methoxy-2-methylpropan-1-amine hydrochloride

  • MDL number MFCD27500929
  • Cas Number : 1803608-40-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-154727

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154727
Availability details for
Quantity
  • Availability
  • Add to Cart