ENA413170303 Enamine

methyl 1-(2-chloroethyl)-1H-1,2,3-triazole-4-carboxylate hydrochloride

  • MDL number MFCD27500931
  • Cas Number : 1803599-99-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154734

Safety Information

msds https://www.enaminestore.com/catalog/EN300-154734
Availability details for
Quantity
  • Availability
  • Add to Cart