ENA413170308 Enamine

1-amino-N-(4-fluorophenyl)cyclopropane-1-carboxamide hydrochloride

  • MDL number MFCD27500936
  • Cas Number : 1803600-13-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154800

Safety Information

msds https://www.enaminestore.com/catalog/EN300-154800
Availability details for
Quantity
  • Availability
  • Add to Cart