ENA413170310 Enamine

2-methyl-4-(1H-pyrrol-3-yl)-1,3-thiazole

  • MDL number MFCD27500938
  • Cas Number : 1803583-12-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154846

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154846
Availability details for
Quantity
  • Availability
  • Add to Cart