ENA413170311 Enamine

2-amino-4-bromo-6-tert-butylphenol

  • MDL number MFCD27500939
  • Cas Number : 1402890-03-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154847

Safety Information

msds https://www.enaminestore.com/catalog/EN300-154847
Availability details for
Quantity
  • Availability
  • Add to Cart