ENA413170317 Enamine

3-methyl-5-phenyl-1H-pyrazol-4-amine

  • MDL number MFCD01825303
  • Cas Number : 112884-51-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152233

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152233
Availability details for
Quantity
  • Availability
  • Add to Cart