ENA413170324 Enamine

2,3-diamino-6-methyl-3,4-dihydropyrimidin-4-one

  • MDL number MFCD00474034
  • Cas Number : 35523-64-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152261

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152261
Availability details for
Quantity
  • Availability
  • Add to Cart