ENA413170325 Enamine

N-(4-fluorophenyl)-5-methyl-1,3,4-thiadiazol-2-amine

  • MDL number MFCD04111436
  • Cas Number : 685113-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 227...229
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152263

Safety Information

msds https://www.enaminestore.com/catalog/EN300-152263
Availability details for
Quantity
  • Availability
  • Add to Cart