ENA413170329 Enamine

1-(3-bromophenyl)-3-cyclopropylpropan-2-amine hydrochloride

  • MDL number MFCD27500735
  • Cas Number : 1798004-11-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152282

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152282
Availability details for
Quantity
  • Availability
  • Add to Cart