ENA413170330 Enamine

3-methylazepane hydrochloride

  • MDL number MFCD27500736
  • Cas Number : 1798010-22-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116...118
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152283

Safety Information

msds https://www.enaminestore.com/catalog/EN300-152283
Availability details for
Quantity
  • Availability
  • Add to Cart