ENA413170339 Enamine

3-fluoro-N-(1H-pyrazol-3-ylmethyl)pyridin-2-amine

  • MDL number MFCD16859505
  • Cas Number : 1343772-56-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152296

Safety Information

msds https://www.enaminestore.com/catalog/EN300-152296
Availability details for
Quantity
  • Availability
  • Add to Cart