ENA413170346 Enamine

1-[2-chloro-4-(4-fluorophenoxy)phenyl]ethan-1-one

  • MDL number MFCD20557802
  • Cas Number : 1021187-74-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152328

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152328
Availability details for
Quantity
  • Availability
  • Add to Cart