ENA413170352 Enamine

7-oxoazepane-4-carboxylic acid

  • MDL number MFCD26936289
  • Cas Number : 38461-79-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152337

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152337
Availability details for
Quantity
  • Availability
  • Add to Cart