ENA413170357 Enamine

3-(2-bromo-4-methylphenyl)propanoic acid

  • MDL number MFCD09999372
  • Cas Number : 829-57-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152376

Safety Information

msds https://www.enaminestore.com/catalog/EN300-152376
Availability details for
Quantity
  • Availability
  • Add to Cart