ENA413170363 Enamine

2-[(2H-indazol-3-yl)formamido]pentanedioic acid

  • MDL number MFCD26936293
  • Cas Number : 1231206-19-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152409

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152409
Availability details for
Quantity
  • Availability
  • Add to Cart