ENA413170364 Enamine

N-methyl-N-propylcarbamoyl chloride

  • MDL number MFCD23790133
  • Cas Number : 51493-02-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152411

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152411
Availability details for
Quantity
  • Availability
  • Add to Cart