ENA413170365 Enamine

methyl 2-(propane-2-sulfonyl)acetate

  • MDL number MFCD12764175
  • Cas Number : 112810-02-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152412

Safety Information

msds https://www.enaminestore.com/catalog/EN300-152412
Availability details for
Quantity
  • Availability
  • Add to Cart