ENA413170370 Enamine

2-chloro-6-(2,2,2-trifluoroethoxy)pyrazine

  • MDL number MFCD09809858
  • Cas Number : 1016530-98-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152423

Safety Information

msds https://www.enaminestore.com/catalog/EN300-152423
Availability details for
Quantity
  • Availability
  • Add to Cart