ENA413170372 Enamine

2-(3,4-diaminophenyl)ethan-1-ol

  • MDL number MFCD18825352
  • Cas Number : 15788-14-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152437

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152437
Availability details for
Quantity
  • Availability
  • Add to Cart