ENA413170385 Enamine

3-(2-fluoro-5-methylphenyl)-1H-pyrazol-5-amine

  • MDL number MFCD16783312
  • Cas Number : 97052-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152454

Safety Information

msds https://www.enaminestore.com/catalog/EN300-152454
Availability details for
Quantity
  • Availability
  • Add to Cart