ENA413170387 Enamine

6-phenyl-2,3-dihydro-1H-inden-1-one

  • MDL number MFCD14654076
  • Cas Number : 170497-64-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152468

Safety Information

msds https://www.enaminestore.com/catalog/EN300-152468
Availability details for
Quantity
  • Availability
  • Add to Cart