ENA413170390 Enamine

N-(2-amino-1-phenylethyl)-3-methoxyaniline

  • MDL number MFCD19355603
  • Cas Number : 1267895-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152477

Safety Information

msds https://www.enaminestore.com/catalog/EN300-152477
Availability details for
Quantity
  • Availability
  • Add to Cart