ENA413170391 Enamine

2-[4-(3-chlorophenyl)piperazin-1-yl]acetonitrile

  • MDL number MFCD01764974
  • Cas Number : 25178-91-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152479

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152479
Availability details for
Quantity
  • Availability
  • Add to Cart