ENA413170395 Enamine

2-[(pyrrolidin-3-yloxy)methyl]-1,3-thiazole

  • MDL number MFCD23873225
  • Cas Number : 933702-08-0
  • Molecular Weight : 184.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-((pyrrolidin-3-yloxy)methyl)thiazole
Inchi Code 1S/C8H12N2OS/c1-2-9-5-7(1)11-6-8-10-3-4-12-8/h3-4,7,9H,1-2,5-6H2
Inchi Key BNHTVWFUKJBIGY-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-152490

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152490
Availability details for
Quantity
  • Availability
  • Add to Cart