ENA413170396 Enamine

4-(2-chlorophenyl)-3,7,7-trimethyl-4H,5H,6H,7H,8H,9H-[1,2]oxazolo[5,4-b]quinolin-5-one

  • MDL number MFCD08486637
  • Cas Number : 852712-88-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152491

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152491
Availability details for
Quantity
  • Availability
  • Add to Cart