ENA413170399 Enamine

2-{4-[1-(methylamino)ethyl]phenyl}-1lambda6,2-thiazolidine-1,1-dione

  • MDL number MFCD12107085
  • Cas Number : 1155097-10-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152496

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152496
Availability details for
Quantity
  • Availability
  • Add to Cart