ENA413170418 Enamine

N-(2,2-dimethylpropyl)aniline

  • MDL number MFCD12774330
  • Cas Number : 7210-81-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152550

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152550
Availability details for
Quantity
  • Availability
  • Add to Cart