ENA413170424 Enamine

N-benzyl-N-methylcarbamoyl chloride

  • MDL number MFCD18431941
  • Cas Number : 32366-02-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152559

Safety Information

msds https://www.enaminestore.com/catalog/EN300-152559
Availability details for
Quantity
  • Availability
  • Add to Cart