ENA413170425 Enamine

4-hydroxy-6,8-dimethoxyquinoline-3-carbonitrile

  • MDL number MFCD13181152
  • Cas Number : 61338-39-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152560

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152560
Availability details for
Quantity
  • Availability
  • Add to Cart