ENA413170427 Enamine

4-({5,7-dimethyl-[1,2,4]triazolo[4,3-a]pyrimidin-3-yl}methyl)aniline

  • MDL number MFCD12589447
  • Cas Number : 1154214-13-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152563

Safety Information

msds https://www.enaminestore.com/catalog/EN300-152563
Availability details for
Quantity
  • Availability
  • Add to Cart